2-Chlorophenylhydrazine hydrochloride, 97%
2-Chlorophenylhydrazine hydrochloride, 97%
2-Chlorophenylhydrazine hydrochloride, 97%
Thermo Scientific Chemicals

2-Chlorophenylhydrazine hydrochloride, 97%

CAS: 41052-75-9 | C6H8Cl2N2 | 179.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 149140050
Price (EUR)
28,10
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
28,10
Each
Chemical Identifiers
CAS41052-75-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light pink to light brown
Infrared spectrumConforms
Titration Argentometric96.0 to 104.0 %
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only