Dimethyl itaconate, 97%, stabilized
Dimethyl itaconate, 97%, stabilized
Dimethyl itaconate, 97%, stabilized
Thermo Scientific Chemicals

Dimethyl itaconate, 97%, stabilized

CAS: 617-52-7 | C7H10O4 | 158.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 149161000
Price (EUR)
35,65
線上優惠
39,10
Save 3,45 (9%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
35,65
線上優惠
39,10
Save 3,45 (9%)
Each
Chemical Identifiers
CAS617-52-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Crystalline mass or liquid
Infrared spectrumConforms
GC>=96.0 %
Melting point34°C to 39°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only