Acetic anhydride, 99+%, pure
Acetic anhydride, 99+%, pure
Acetic anhydride, 99+%, pure
Thermo Scientific Chemicals

Acetic anhydride, 99+%, pure

CAS: 108-24-7 | C4H6O3 | 102.089 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
25 L
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 149490025
Price (EUR)
-
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS108-24-7
IUPAC Nameacetyl acetate
Molecular FormulaC4H6O3
InChI KeyWFDIJRYMOXRFFG-UHFFFAOYSA-N
SMILESCC(=O)OC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Refractive index1.3890 to 1.3910 (20°C, 589 nm)
Infrared spectrumConforms
GC>=99.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent
RUO – Research Use Only