Valeraldehyde, 97%
Valeraldehyde, 97%
Valeraldehyde, 97%
Thermo Scientific Chemicals

Valeraldehyde, 97%

CAS: 110-62-3 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 149560010
Price (EUR)
94,40
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
94,40
Each
Chemical Identifiers
CAS110-62-3
IUPAC Namepentanal
Molecular FormulaC5H10O
InChI KeyHGBOYTHUEUWSSQ-UHFFFAOYSA-N
SMILESCCCCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Infrared spectrumConforms
Refractive index1.3925 to 1.3975 (20°C, 589 nm)
Free acid=<1.5 %
Appearance (Color)Clear colorless to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only