3,4-Dichlorobenzoyl chloride, 97%
3,4-Dichlorobenzoyl chloride, 97%
3,4-Dichlorobenzoyl chloride, 97%
Thermo Scientific Chemicals

3,4-Dichlorobenzoyl chloride, 97%

CAS: 3024-72-4 | C7H3Cl3O | 209.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Catalog number 150230050
Price (EUR)
18,30
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
18,30
Each
Chemical Identifiers
CAS3024-72-4
IUPAC Name3,4-dichlorobenzoyl chloride
Molecular FormulaC7H3Cl3O
InChI KeyVTXNOVCTHUBABW-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting crystalline solid
Infrared spectrumConforms
Titration RCOCl>=96.0 %
GC>=96.0 %
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only