Bromocresol Purple, pure, indicator grade
Bromocresol Purple, pure, indicator grade
Bromocresol Purple, pure, indicator grade
Thermo Scientific Chemicals

Bromocresol Purple, pure, indicator grade

CAS: 115-40-2 | C21H16Br2O5S | 540.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
140 g
1 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 151331400
Price (EUR)
391,00
Each
Quantity:
140 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
391,00
Each
Chemical Identifiers
CAS115-40-2
IUPAC Name3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC21H16Br2O5S
InChI KeyABIUHPWEYMSGSR-UHFFFAOYSA-N
SMILESCC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light pink to pink-orange
UV(20 mg/l in methanol) Complies
Infrared spectrumConforms
Solubility(0.1 % in 50 % ethanol) Clear
Visual transition intervalfrom pH 5.2 (yellow) to pH 6.8 (purple)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only