Cresol Red, sodium salt, pure, water soluble, indicator
Cresol Red, sodium salt, pure, water soluble, indicator
Cresol Red, sodium salt, pure, water soluble, indicator
Thermo Scientific Chemicals

Cresol Red, sodium salt, pure, water soluble, indicator

CAS: 62625-29-0 | C21H17NaO5S
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 151390250
Price (EUR)
71,90
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
71,90
Each
Chemical Identifiers
CAS62625-29-0
IUPAC Namesodium 4-[3-(4-hydroxy-3-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-2-methylbenzen-1-olate
Molecular FormulaC21H17NaO5S
InChI KeyFMKJVYROEYMYFV-UHFFFAOYNA-M
SMILES[Na+].CC1=CC(=CC=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C([O-])C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Visual transition intervalfrom pH 7.0 (yellow) to pH 8.8 (purple)
Appearance (Color)Brown
Solubility(0.1 % in water) Clear
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only