5-(4-Dimethylaminobenzylidene)rhodanine, 99%
5-(4-Dimethylaminobenzylidene)rhodanine, 99%
5-(4-Dimethylaminobenzylidene)rhodanine, 99%
Thermo Scientific Chemicals

5-(4-Dimethylaminobenzylidene)rhodanine, 99%

5-(4-Dimethylaminobenzylidene)rhodanine, 99%, Quantity: 100g, Packaging: Glass bottle, Red, Melting Point: 285 deg.C to 288 deg.C, Molecular Weight: 264.35, Percent Purity: 99%, Appearance: (0.3% soln. in acetone) clear, Assay Percent Range: 99%, Beilstein: 27, 433, CAS: 536-17-4 | CAS: 536-17-4 | C12H12N2OS2 | 264.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 151470250
Price (EUR)
113,65
線上優惠
126,00
Save 12,35 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
113,65
線上優惠
126,00
Save 12,35 (10%)
Each
Chemical Identifiers
CAS536-17-4
IUPAC Name(5Z)-5-{[4-(dimethylamino)phenyl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular FormulaC12H12N2OS2
InChI KeyJJRVRELEASDUMY-YFHOEESVSA-N
SMILESCN(C)C1=CC=C(C=C2/SC(=S)NC2=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with TBAH98.5 to 101.5 %
Suitability(for Ag determination) Passes test
Infrared spectrumConforms
Appearance (Form)Powder
Solubility(0.03 % in acetone) Clear
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only