4-Chromanone, 96%
4-Chromanone, 96%
4-Chromanone, 96%
Thermo Scientific Chemicals

4-Chromanone, 96%

CAS: 491-37-2 | C9H8O2 | 148.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 153460500
Price (EUR)
256,00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
256,00
Each
Chemical Identifiers
CAS491-37-2
IUPAC Name3,4-dihydro-2H-1-benzopyran-4-one
Molecular FormulaC9H8O2
InChI KeyMSTDXOZUKAQDRL-UHFFFAOYSA-N
SMILESO=C1CCOC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Appearance (Form)Crystalline powder or crystals or low melting mass
Infrared spectrumConforms
Melting point34°C to 40°C
GC>=95 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only