Alizarin, 97%, pure
Alizarin, 97%, pure
Alizarin, 97%, pure
Thermo Scientific Chemicals

Alizarin, 97%, pure

CAS: 72-48-0 | C14H8O4 | 240.214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 153695000
Price (EUR)
385,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
385,00
Each
Chemical Identifiers
CAS72-48-0
IUPAC Name1,2-dihydroxy-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H8O4
InChI KeyRGCKGOZRHPZPFP-UHFFFAOYSA-N
SMILESOC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Dye content>=96.0 %
Appearance (Color)Orange to orange-brown
Loss on drying=<2 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only