chlorodiphenylmethane, 98%
chlorodiphenylmethane, 98%
chlorodiphenylmethane, 98%
Thermo Scientific Chemicals

chlorodiphenylmethane, 98%

CAS: 90-99-3 | C13H11Cl | 202.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 154660050
Price (EUR)
22,80
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
22,80
Each
Chemical Identifiers
CAS90-99-3
IUPAC Name[chloro(phenyl)methyl]benzene
Molecular FormulaC13H11Cl
InChI KeyZDVDCDLBOLSVGM-UHFFFAOYSA-N
SMILESClC(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow
GC>=97.5 %
Appearance (Form)Liquid
Refractive index1.5940 to 1.5970 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only