Citraconic anhydride, 98%
Citraconic anhydride, 98%
Citraconic anhydride, 98%
Citraconic anhydride, 98%
Thermo Scientific Chemicals

Citraconic anhydride, 98%

CAS: 616-02-4 | C5H4O3 | 112.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Catalog number 154790250
Price (EUR)
26,10
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
26,10
Each
Add to cart
Chemical Identifiers
CAS616-02-4
IUPAC Name3-methyl-2,5-dihydrofuran-2,5-dione
Molecular FormulaC5H4O3
InChI KeyAYKYXWQEBUNJCN-UHFFFAOYSA-N
SMILESCC1=CC(=O)OC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Titration with NaOH97.5 to 102.5 %
Refractive index1.4690 to 1.4730 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only