2'-Deoxyadenosine hydrate, 99%
2'-Deoxyadenosine hydrate, 99%
2'-Deoxyadenosine hydrate, 99%
Thermo Scientific Chemicals

2'-Deoxyadenosine hydrate, 99%

CAS: 16373-93-6 | C10H13N5O3 | 251.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 154990050
Price (EUR)
138,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
138,00
Each
Chemical Identifiers
CAS16373-93-6
IUPAC Name(2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-2-(hydroxymethyl)oxolan-3-ol
Molecular FormulaC10H13N5O3
InChI KeyOLXZPDWKRNYJJZ-YUZWJPFSNA-N
SMILESNC1=C2N=CN([C@H]3C[C@H](O)[C@@H](CO)O3)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=98.5 %
Water=<8 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only