2,3-Diaminopyridine, 98%
2,3-Diaminopyridine, 98%
2,3-Diaminopyridine, 98%
Thermo Scientific Chemicals

2,3-Diaminopyridine, 98%

CAS: 452-58-4 | C5H7N3 | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
25 g
Packaging:
Glass bottle
Catalog number 155020050
Price (EUR)
77,80
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
77,80
Each
Chemical Identifiers
CAS452-58-4
IUPAC Namepyridine-2,3-diamine
Molecular FormulaC5H7N3
InChI KeyZZYXNRREDYWPLN-UHFFFAOYSA-N
SMILESNC1=CC=CN=C1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or needles
GC>=97.5 %
Infrared spectrumConforms
Appearance (Color)Grey to dark brown
AppearanceMay darken on storage
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only