4,5-Dihydroxynaphthalene-2,7-disulfonic Acid, Disodium Salt Dihydrate, 98%
4,5-Dihydroxynaphthalene-2,7-disulfonic Acid, Disodium Salt Dihydrate, 98%
4,5-Dihydroxynaphthalene-2,7-disulfonic Acid, Disodium Salt Dihydrate, 98%
Thermo Scientific Chemicals

4,5-Dihydroxynaphthalene-2,7-disulfonic Acid, Disodium Salt Dihydrate, 98%

4,5-Dihydroxynaphthalene-2,7-disulfonic acid, disodium salt dihydrate, 98%, C10H6Na2O8S2.2H2O, CAS Number-5808-22-0 | CAS: 5808-22-0 | C10H6O8S2 | 318.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 155195000
Price (EUR)
342,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
342,00
Each
Chemical Identifiers
CAS5808-22-0
IUPAC Name2,7-disodium 4,5-dihydroxynaphthalene-2,7-disulfonate dihydrate
Molecular FormulaC10H10Na2O10S2
InChI KeyQUEAKWJKJBFNEG-UHFFFAOYSA-L
SMILESO.O.OC1=CC(=CC2=CC(=CC(O)=C12)S(=O)(=O)O[Na])S(=O)(=O)O[Na]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Appearance (Color)Off-white to beige to grey to light brown
Suitability(for det. of NO3 and HCHO) Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only