1,3-Dimethoxybenzene, 99%
1,3-Dimethoxybenzene, 99%
1,3-Dimethoxybenzene, 99%
Thermo Scientific Chemicals

1,3-Dimethoxybenzene, 99%

CAS: 151-10-0 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 155231000
Price (EUR)
33,30
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
33,30
Each
Chemical Identifiers
CAS151-10-0
IUPAC Name1,3-dimethoxybenzene
Molecular FormulaC8H10O2
InChI KeyDPZNOMCNRMUKPS-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.5230 to 1.5250 (20°C, 589 nm)
Appearance (Color)Clear colorless to light brown
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only