Dimethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Dimethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Dimethylamine hydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethylamine hydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
1 kg
2.5 kg
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 155270025
Price (EUR)
-
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS506-59-2
IUPAC Namehydrogen dimethylamine chloride
Molecular FormulaC2H8ClN
InChI KeyIQDGSYLLQPDQDV-UHFFFAOYSA-N
SMILES[H+].[Cl-].CNC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
pH5 to 6 (5 % in water)
Appearance (Color)White to almost white
Titration Argentometric98.5 to 101.5 % (on dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only