N-Ethylmaleimide, 98+%
N-Ethylmaleimide, 98+%
N-Ethylmaleimide, 98+%
Thermo Scientific Chemicals

N-Ethylmaleimide, 98+%

CAS: 128-53-0 | C6H7NO2 | 125.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
10 g
50 g
250 g
Catalog number 156100500
Price (EUR)
524,00
Each
Quantity:
50 g
Request bulk or custom format
Price (EUR)
524,00
Each
Chemical Identifiers
CAS128-53-0
IUPAC Name1-ethyl-2,5-dihydro-1H-pyrrole-2,5-dione
Molecular FormulaC6H7NO2
InChI KeyHDFGOPSGAURCEO-UHFFFAOYSA-N
SMILESCCN1C(=O)C=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Solubility(5% in 100% EtOH) Clear to light turbid,
SolubilityColorless to light yellow solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only