3-Methyl-3-buten-1-ol, 97%
3-Methyl-3-buten-1-ol, 97%
3-Methyl-3-buten-1-ol, 97%
Thermo Scientific Chemicals

3-Methyl-3-buten-1-ol, 97%

CAS: 763-32-6 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 156635000
Price (EUR)
121,65
Online Exclusive
135,00
Save 13,35 (10%)
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
121,65
Online Exclusive
135,00
Save 13,35 (10%)
Each
Chemical Identifiers
CAS763-32-6
IUPAC Name3-methylbut-3-en-1-ol
Molecular FormulaC5H10O
InChI KeyCPJRRXSHAYUTGL-UHFFFAOYSA-N
SMILESCC(=C)CCO
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4327 to 1.4347 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only