1-Methylpiperazine, 99%
1-Methylpiperazine, 99%
1-Methylpiperazine, 99%
Thermo Scientific Chemicals

1-Methylpiperazine, 99%

CAS: 109-01-3 | C5H12N2 | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
View all
Metal Drum
Catalog number 156910025
Price (EUR)
546,00
Each
Quantity:
2.5 kg
Packaging:
Metal Drum
Request bulk or custom format
Price (EUR)
546,00
Each
Chemical Identifiers
CAS109-01-3
IUPAC Name1-methylpiperazine
Molecular FormulaC5H12N2
InChI KeyPVOAHINGSUIXLS-UHFFFAOYSA-N
SMILESCN1CCNCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<20 APHA
Infrared spectrumConforms
Refractive index1.4645 to 1.4665 (20°C, 589 nm)
GC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only