2-Nitrophenylacetic acid, 99%
2-Nitrophenylacetic acid, 99%
2-Nitrophenylacetic acid, 99%
Thermo Scientific Chemicals

2-Nitrophenylacetic acid, 99%

CAS: 3740-52-1 | C8H7NO4 | 181.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 157060250
Price (EUR)
51,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
51,00
Each
Chemical Identifiers
CAS3740-52-1
IUPAC Name2-(2-nitrophenyl)acetic acid
Molecular FormulaC8H7NO4
InChI KeyWMUZDBZPDLHUMW-UHFFFAOYSA-N
SMILESOC(=O)CC1=CC=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point138°C to 142°C
Appearance (Form)Crystalline powder
HPLC>=98.5 %
Appearance (Color)Yellow to beige
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only