Tris(2-butoxyethyl) phosphate, 95%
Tris(2-butoxyethyl) phosphate, 95%
Tris(2-butoxyethyl) phosphate, 95%
Thermo Scientific Chemicals

Tris(2-butoxyethyl) phosphate, 95%

CAS: 78-51-3 | C18H39O7P | 398.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 157240050
Price (EUR)
19,65
Online exclusive
21,40
Save 1,75 (8%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
19,65
Online exclusive
21,40
Save 1,75 (8%)
Each
Chemical Identifiers
CAS78-51-3
IUPAC Nametris(2-butoxyethyl) phosphate
Molecular FormulaC18H39O7P
InChI KeyWTLBZVNBAKMVDP-UHFFFAOYSA-N
SMILESCCCCOCCOP(=O)(OCCOCCCC)OCCOCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=94 %
Appearance (Form)Liquid
Infrared spectrumConforms
Refractive index1.4360 to 1.4390 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only