DL-4-Chlorophenylalanine, 98%
DL-4-Chlorophenylalanine, 98%
DL-4-Chlorophenylalanine, 98%
Thermo Scientific Chemicals

DL-4-Chlorophenylalanine, 98%

CAS: 7424-00-2 | C9H10ClNO2 | 199.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
View all
Plastic Bottle
Catalog number 157280250
Price (EUR)
215,65
Online Exclusive
239,00
Save 23,35 (10%)
Each
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (EUR)
215,65
Online Exclusive
239,00
Save 23,35 (10%)
Each
Chemical Identifiers
CAS7424-00-2
IUPAC Name2-amino-3-(4-chlorophenyl)propanoic acid
Molecular FormulaC9H10ClNO2
InChI KeyNIGWMJHCCYYCSF-UHFFFAOYNA-N
SMILESNC(CC1=CC=C(Cl)C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only