Triethylamine, 99%, pure
Triethylamine, 99%, pure
Triethylamine, 99%, pure
Thermo Scientific Chemicals

Triethylamine, 99%, pure

CAS: 121-44-8 | C6H15N | 101.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 157910050
Price (EUR)
28,65
キャンペーン価格
32,00
Save 3,35 (10%)
Each
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
28,65
キャンペーン価格
32,00
Save 3,35 (10%)
Each
Chemical Identifiers
CAS121-44-8
IUPAC Nametriethylamine
Molecular FormulaC6H15N
InChI KeyZMANZCXQSJIPKH-UHFFFAOYSA-N
SMILESCCN(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<15 APHA
Infrared spectrumConforms
GC>=98.5 %
Water=<0.2 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only