Ethyl bromoacetate, 98%
Ethyl bromoacetate, 98%
Ethyl bromoacetate, 98%
Thermo Scientific Chemicals

Ethyl bromoacetate, 98%

CAS: 105-36-2 | C4H7BrO2 | 167 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 158591000
Price (EUR)
32,30
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
32,30
Each
Chemical Identifiers
CAS105-36-2
IUPAC Nameethyl 2-bromoacetate
Molecular FormulaC4H7BrO2
InChI KeyPQJJJMRNHATNKG-UHFFFAOYSA-N
SMILESCCOC(=O)CBr
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Infrared spectrumConforms
Color scale=<300 APHA
Refractive index1.4500 to 1.4520 (20°C, 589 nm)
Appearance (Form)Clear liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only