Morpholine, 99+%, extra pure
Morpholine, 99+%, extra pure
Morpholine, 99+%, extra pure
Thermo Scientific Chemicals

Morpholine, 99+%, extra pure

Morpholine, 99+%, C4H9NO, CAS Number-110-91-8, 1-oxa-4-azacyclohexane, diethyleneimide oxide, diethylene imidoxide, diethylene oximide, diethylenimide oxide, drewamine, p-isoxazine, tetrahydro, tetrahydro-1,4-oxazine, tetrahydro-2h-1,4-oxazine, tetrahydro-p-oxazine, 1L, 99% min. (GC), 27, 5 | CAS: 110-91-8 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
1 L
2.5 L
10 L
Packaging:
View all
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 158680025
Price (EUR)
67,30
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
67,30
Each
Chemical Identifiers
CAS110-91-8
IUPAC Namemorpholine
Molecular FormulaC4H9NO
InChI KeyYNAVUWVOSKDBBP-UHFFFAOYSA-N
SMILESC1COCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Clear liquid
GC>=99.0 %
Color scale=<20 APHA
Refractive index1.4530 to 1.4550 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only