Vinylacetic acid, tech., 90%, unstabilized
Vinylacetic acid, tech., 90%, unstabilized
Vinylacetic acid, tech., 90%, unstabilized
Thermo Scientific Chemicals

Vinylacetic acid, tech., 90%, unstabilized

CAS: 625-38-7 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 158830250
Price (EUR)
99,30
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
99,30
Each
Chemical Identifiers
CAS625-38-7
IUPAC Namebut-3-enoic acid
Molecular FormulaC4H6O2
InChI KeyPVEOYINWKBTPIZ-UHFFFAOYSA-N
SMILESOC(=O)CC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=88 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only