Catechol, 99+%
Catechol, 99+%
Catechol, 99+%
Catechol, 99+%
Thermo Scientific Chemicals

Catechol, 99+%

CAS: 120-80-9 | C6H6O2 | 110.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 158981000
Price (EUR)
34,65
キャンペーン価格
38,70
Save 4,05 (10%)
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
34,65
キャンペーン価格
38,70
Save 4,05 (10%)
Each
Add to cart
Chemical Identifiers
CAS120-80-9
IUPAC Namebenzene-1,2-diol
Molecular FormulaC6H6O2
InChI KeyYCIMNLLNPGFGHC-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline flakes
Melting point103°C to 106°C
AppearanceMay darken on storage
Infrared spectrumConforms
Appearance (Color)Off-white to grey-brown
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only