Cholic acid, 97%
Cholic acid, 97%
Cholic acid, 97%
Thermo Scientific Chemicals

Cholic acid, 97%

CAS: 81-25-4 | C24H40O5 | 408.579 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 159115000
Price (EUR)
317,00
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
317,00
Each
Chemical Identifiers
CAS81-25-4
IUPAC Name(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Molecular FormulaC24H40O5
InChI KeyBHQCQFFYRZLCQQ-OELDTZBJSA-N
SMILESC[C@H](CCC(O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy metals=<20 ppm
Specific optical rotation(20°C, 589 nm) (c=2, ethanol) > +35°
Titration with NaOH>=96.0 %
Appearance (Form)Crystalline powder
Melting point197°C to 202°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only