2-Chlorobenzoic acid, 98%
2-Chlorobenzoic acid, 98%
2-Chlorobenzoic acid, 98%
Thermo Scientific Chemicals

2-Chlorobenzoic acid, 98%

CAS: 118-91-2 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2 kg
Packaging:
Plastic bottle
Catalog number 159240020
Price (EUR)
140,00
Each
Quantity:
2 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
140,00
Each
Chemical Identifiers
CAS118-91-2
IUPAC Name2-chlorobenzoic acid
Molecular FormulaC7H5ClO2
InChI KeyIKCLCGXPQILATA-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
HPLC>=97.5 %
Appearance (Form)Powder
Melting point139°C to 143°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only