2,3-Diaminonaphthalene, 97%
2,3-Diaminonaphthalene, 97%
2,3-Diaminonaphthalene, 97%
Thermo Scientific Chemicals

2,3-Diaminonaphthalene, 97%

CAS: 771-97-1 | C10H10N2 | 158.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 159590050
Price (EUR)
770,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
770,00
Each
Chemical Identifiers
CAS771-97-1
IUPAC Namenaphthalene-2,3-diamine
Molecular FormulaC10H10N2
InChI KeyXTBLDMQMUSHDEN-UHFFFAOYSA-N
SMILESNC1=C(N)C=C2C=CC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green-brown to brown
Appearance (Form)Powder
HPLC>=96.0 %
Infrared spectrumConforms
Melting point193°C to 199°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only