3-Chlorotoluene, 97%
3-Chlorotoluene, 97%
3-Chlorotoluene, 97%
Thermo Scientific Chemicals

3-Chlorotoluene, 97%

CAS: 108-41-8 | C7H7Cl | 126.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
500 mL
Packaging:
Glass bottle
Catalog number 160451000
Price (EUR)
42,90
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
42,90
Each
Chemical Identifiers
CAS108-41-8
IUPAC Name1-chloro-3-methylbenzene
Molecular FormulaC7H7Cl
InChI KeyOSOUNOBYRMOXQQ-UHFFFAOYSA-N
SMILESCC1=CC=CC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=96.0 %
Refractive index1.5210 to 1.5230 (20°C, 589 nm)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only