Phenyltrimethylammonium tribromide, 97%
Phenyltrimethylammonium tribromide, 97%
Phenyltrimethylammonium tribromide, 97%
Thermo Scientific Chemicals

Phenyltrimethylammonium tribromide, 97%

CAS: 4207-56-1 | C9H14Br3N | 375.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 160940250
Price (EUR)
43,50
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
43,50
Each
Chemical Identifiers
CAS4207-56-1
IUPAC NameN,N,N-trimethylanilinium; tribroman-2-uide
Molecular FormulaC9H14Br3N
InChI KeyPRXNKYBFWAWBNZ-UHFFFAOYSA-N
SMILESBr[Br-]Br.C[N+](C)(C)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point113°C to 117°C
Titration>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only