4,4'-Dimethoxybenzophenone, 97%
4,4'-Dimethoxybenzophenone, 97%
4,4'-Dimethoxybenzophenone, 97%
Thermo Scientific Chemicals

4,4'-Dimethoxybenzophenone, 97%

CAS: 90-96-0 | C15H14O3 | 242.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 161740250
Price (EUR)
68,20
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
68,20
Each
Chemical Identifiers
CAS90-96-0
IUPAC Namebis(4-methoxyphenyl)methanone
Molecular FormulaC15H14O3
InChI KeyRFVHVYKVRGKLNK-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1)C(=O)C1=CC=C(OC)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to light beige
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Form)Powder
Melting point141°C to 146°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only