3-Methyl-2-cyclopenten-1-one, 98%, stabilized
3-Methyl-2-cyclopenten-1-one, 98%, stabilized
3-Methyl-2-cyclopenten-1-one, 98%, stabilized
Thermo Scientific Chemicals

3-Methyl-2-cyclopenten-1-one, 98%, stabilized

CAS: 2758-18-1 | C6H8O | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
View all
Glass bottle
Catalog number 163590100
Price (EUR)
72,10
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
72,10
Each
Chemical Identifiers
CAS2758-18-1
IUPAC Name3-methylcyclopent-2-en-1-one
Molecular FormulaC6H8O
InChI KeyCHCCBPDEADMNCI-UHFFFAOYSA-N
SMILESCC1=CC(=O)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Stabilizer0.1 % hydroquinone
Appearance (Color)Clear light yellow to yellow-brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only