3-Methylbutyraldehyde, 98%
3-Methylbutyraldehyde, 98%
3-Methylbutyraldehyde, 98%
Thermo Scientific Chemicals

3-Methylbutyraldehyde, 98%

CAS: 590-86-3 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 163975000
Price (EUR)
60,40
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
60,40
Each
Chemical Identifiers
CAS590-86-3
IUPAC Name3-methylbutanal
Molecular FormulaC5H10O
InChI KeyYGHRJJRRZDOVPD-UHFFFAOYSA-N
SMILESCC(C)CC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
Refractive index1.3870 to 1.3890 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only