2-Indanone, 97%
2-Indanone, 97%
2-Indanone, 97%
Thermo Scientific Chemicals

2-Indanone, 97%

CAS: 615-13-4 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 164110100
Price (EUR)
80,40
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
80,40
Each
Chemical Identifiers
CAS615-13-4
IUPAC Name2,3-dihydro-1H-inden-2-one
Molecular FormulaC9H8O
InChI KeyUMJJFEIKYGFCAT-UHFFFAOYSA-N
SMILESO=C1CC2=CC=CC=C2C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to brown
Appearance (Form)Powder or crystals and/or chunks
GC>=96.0 %
Melting point50°C to 57°C
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only