4,4'-Bis(dimethylamino)benzophenone, 98%
4,4'-Bis(dimethylamino)benzophenone, 98%
4,4'-Bis(dimethylamino)benzophenone, 98%
Thermo Scientific Chemicals

4,4'-Bis(dimethylamino)benzophenone, 98%

CAS: 90-94-8 | C17H20N2O | 268.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 164490050
Price (EUR)
23,90
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
23,90
Each
Chemical Identifiers
CAS90-94-8
IUPAC Name4-[4-(dimethylamino)benzoyl]-N,N-dimethylaniline
Molecular FormulaC17H20N2O
InChI KeyVVBLNCFGVYUYGU-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=C1)C(=O)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue to dark blue to grey
Appearance (Form)Powder
Infrared spectrumConforms
Melting point171°C to 176°C (Dec)
Titration with HClO497.5 to 102.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only