Pyrogallol, 99%
Pyrogallol, 99%
Pyrogallol, 99%
Thermo Scientific Chemicals

Pyrogallol, 99%

CAS: 87-66-1 | C6H6O3 | 126.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 164565000
Price (EUR)
318,65
Precio exclusivo compra web
354,00
Save 35,35 (10%)
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
318,65
Precio exclusivo compra web
354,00
Save 35,35 (10%)
Each
Chemical Identifiers
CAS87-66-1
IUPAC Namebenzene-1,2,3-triol
Molecular FormulaC6H6O3
InChI KeyWQGWDDDVZFFDIG-UHFFFAOYSA-N
SMILESOC1=CC=CC(O)=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfated ash=<0.1 %
Infrared spectrumConforms
Appearance (Color)White to off-white
HPLC>=98.5 %
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only