N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis
N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis
N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis
Thermo Scientific Chemicals

N,N'-Methylenebisacrylamide, 99+%, for biochemistry, for electrophoresis

CAS: 110-26-9 | C7H10N2O2 | 154.169 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
Catalog number 164791000
Price (EUR)
183,00
Each
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (EUR)
183,00
Each
Chemical Identifiers
CAS110-26-9
IUPAC NameN-[(prop-2-enamido)methyl]prop-2-enamide
Molecular FormulaC7H10N2O2
InChI KeyZIUHHBKFKCYYJD-UHFFFAOYSA-N
SMILESC=CC(=O)NCNC(=O)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Infrared spectrumConforms
UV(1 % in water) 1 cm cell
Appearance (Form)Crystalline powder or crystals or needles
Acrylic acid=<0.05 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only