5-Nitro-2-furaldehyde diacetate, 98%
5-Nitro-2-furaldehyde diacetate, 98%
5-Nitro-2-furaldehyde diacetate, 98%
Thermo Scientific Chemicals

5-Nitro-2-furaldehyde diacetate, 98%

CAS: 92-55-7 | C9H9NO7 | 243.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 165131000
Price (EUR)
32,30
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
32,30
Each
Chemical Identifiers
CAS92-55-7
IUPAC Name(acetyloxy)(5-nitrofuran-2-yl)methyl acetate
Molecular FormulaC9H9NO7
InChI KeyHSXKWKJCZNRMJO-UHFFFAOYSA-N
SMILESCC(=O)OC(OC(C)=O)C1=CC=C(O1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder
GC>=97.5 %
Melting point89°C to 93°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only