Potassium tert-butoxide, 98+%, pure
Potassium tert-butoxide, 98+%, pure
Potassium tert-butoxide, 98+%, pure
Thermo Scientific Chemicals

Potassium tert-butoxide, 98+%, pure

CAS: 865-47-4 | C4H9KO | 112.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
Catalog number 168880025
Price (EUR)
548,65
Online exclusive
609,00
Save 60,35
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
548,65
Online exclusive
609,00
Save 60,35
Each
Chemical Identifiers
CAS865-47-4
IUPAC Namepotassium 2-methylpropan-2-olate
Molecular FormulaC4H9KO
InChI KeyLPNYRYFBWFDTMA-UHFFFAOYSA-N
SMILES[K+].CC(C)(C)[O-]
View more
SpecificationsSpecification SheetSpecification Sheet
KOH + K2CO3 (as KOH)=<0.50 %
Appearance (Form)Powder
Appearance (Color)White to off-white
Titration with HCl>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only