4-tert-Butylbenzoyl chloride, 98%
4-tert-Butylbenzoyl chloride, 98%
4-tert-Butylbenzoyl chloride, 98%
Thermo Scientific Chemicals

4-tert-Butylbenzoyl chloride, 98%

CAS: 1710-98-1 | C11H13ClO | 196.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 169100250
Price (EUR)
62,40
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
62,40
Each
Chemical Identifiers
CAS1710-98-1
IUPAC Name4-tert-butylbenzoyl chloride
Molecular FormulaC11H13ClO
InChI KeyWNLMYNASWOULQY-UHFFFAOYSA-N
SMILESCC(C)(C)C1=CC=C(C=C1)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Titration RCOCl>=97.5 %
Refractive index1.5354 to 1.5374 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only