2-Bromoanisole, 97%
2-Bromoanisole, 97%
2-Bromoanisole, 97%
Thermo Scientific Chemicals

2-Bromoanisole, 97%

CAS: 578-57-4 | C7H7BrO | 187.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 170260250
Price (EUR)
59,80
Each
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
59,80
Each
Add to cart
Chemical Identifiers
CAS578-57-4
IUPAC Name1-bromo-2-methoxybenzene
Molecular FormulaC7H7BrO
InChI KeyHTDQSWDEWGSAMN-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=96.0 %
Infrared spectrumConforms
Refractive index1.5730 to 1.5750 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only