alpha-Bromo-p-toluic acid, 97%
alpha-Bromo-p-toluic acid, 97%
alpha-Bromo-p-toluic acid, 97%
Thermo Scientific Chemicals

alpha-Bromo-p-toluic acid, 97%

CAS: 6232-88-8 | C8H7BrO2 | 215.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
View all
Glass Bottle
Catalog number 170402500
Price (EUR)
-
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS6232-88-8
IUPAC Name4-(bromomethyl)benzoic acid
Molecular FormulaC8H7BrO2
InChI KeyCQQSQBRPAJSTFB-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(CBr)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=96.0 %
HPLC>=96.0 %
Appearance (Color)White to beige
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only