2-Hydroxy-4-methoxybenzaldehyde, 99%
2-Hydroxy-4-methoxybenzaldehyde, 99%
2-Hydroxy-4-methoxybenzaldehyde, 99%
Thermo Scientific Chemicals

2-Hydroxy-4-methoxybenzaldehyde, 99%

CAS: 673-22-3 | C8H8O3 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 171080050
Price (EUR)
85,20
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
85,20
Each
Chemical Identifiers
CAS673-22-3
IUPAC Name2-hydroxy-4-methoxybenzaldehyde
Molecular FormulaC8H8O3
InChI KeyWZUODJNEIXSNEU-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to beige to light brown
Infrared spectrumConforms
Melting point40°C to 43°C
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only