2-(2-Chloroethoxy)ethanol, 99%
2-(2-Chloroethoxy)ethanol, 99%
2-(2-Chloroethoxy)ethanol, 99%
Thermo Scientific Chemicals

2-(2-Chloroethoxy)ethanol, 99%

CAS: 628-89-7 | C4H9ClO2 | 124.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 172260500
Price (EUR)
48,50
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
48,50
Each
Chemical Identifiers
CAS628-89-7
IUPAC Name2-(2-chloroethoxy)ethan-1-ol
Molecular FormulaC4H9ClO2
InChI KeyLECMBPWEOVZHKN-UHFFFAOYSA-N
SMILESOCCOCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Water=<1 % (K.F.)
Color scale=<20 APHA
GC>=98.5 %
Refractive index1.4519 to 1.4539 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only