4-Amino-5-imidazolecarboxamide hydrochloride, 98%
4-Amino-5-imidazolecarboxamide hydrochloride, 98%
4-Amino-5-imidazolecarboxamide hydrochloride, 98%
Thermo Scientific Chemicals

4-Amino-5-imidazolecarboxamide hydrochloride, 98%

CAS: 72-40-2 | C4H7ClN4O | 162.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 173090250
Price (EUR)
203,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
203,00
Each
Chemical Identifiers
CAS72-40-2
IUPAC Name4-amino-1H-imidazole-5-carboxamide hydrochloride
Molecular FormulaC4H7ClN4O
InChI KeyMXCUYSMIELHIQL-UHFFFAOYSA-N
SMILESCl.NC(=O)C1=C(N)N=CN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige to light grey
Titration Argentometric>=97.5 %
Infrared spectrumConforms
Solubility(5 % in water) Clear
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only