Sulfobromophthalein sodium hydrate
Sulfobromophthalein sodium hydrate
Sulfobromophthalein sodium hydrate
Thermo Scientific Chemicals

Sulfobromophthalein sodium hydrate

CAS: 123359-42-2 | C20H8Br4Na2O10S2 | 837.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 173400050
Price (EUR)
297,00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
297,00
Each
Chemical Identifiers
CAS123359-42-2
IUPAC Namedisodium 2-hydroxy-5-[4,5,6,7-tetrabromo-1-(4-hydroxy-3-sulfonatophenyl)-3-oxo-1,3-dihydro-2-benzofuran-1-yl]benzene-1-sulfonate
Molecular FormulaC20H8Br4Na2O10S2
InChI KeyGHAFORRTMVIXHS-UHFFFAOYSA-L
SMILES[Na+].[Na+].OC1=CC=C(C=C1S([O-])(=O)=O)C1(OC(=O)C2=C1C(Br)=C(Br)C(Br)=C2Br)C1=CC=C(O)C(=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white to grey
UV Lambda max575 to 581 nm (c=10 mg/l, 0.1N NaOH)
Infrared spectrumConforms
HPLC>=95 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only