Chloroacetone, 96%, stabilized
Chloroacetone, 96%, stabilized
Chloroacetone, 96%, stabilized
Thermo Scientific Chemicals

Chloroacetone, 96%, stabilized

CAS: 78-95-5 | C3H5ClO | 92.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
250 mL
1 L
5 L
Packaging:
View all
Glass bottle
Catalog number 173470250
Price (EUR)
21,50
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
21,50
Each
Chemical Identifiers
CAS78-95-5
IUPAC Name1-chloropropan-2-one
Molecular FormulaC3H5ClO
InChI KeyBULLHNJGPPOUOX-UHFFFAOYSA-N
SMILESCC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=95 %
Refractive index1.4310 to 1.4370 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only